Learn More
Heinz Herenz Wooden Applicator With Cellulose Bud Single End Tip
Product Code. 30158461
Click to view available options
Tip Type:
Single End
Unit Size:
Pack of 100
Product Code. 30158461
missing translation for 'mfr': Heinz Herenz 1032518
Wooden applicator WA 11. Thin with cellulose bud.
We offer a wide range of sterile and nonsterile swabs:
  • Wood, PS, PP, aluminium or paper applicator
  • Each in various lengths available
  • With cotton, viscose, polyester or foam head
  • Diameter of the heads: small, medium or large
  • This guarantees that the right instrument is available for every special application.
  • For example, the cotton-tipped swabs are recommended for use by ENT specialists.
  • Our sterile cotton buds are all individually wrapped.
Please note in the medical field:
  • Only suitable for single use
  • Use by medical personnel only
Specifications
Autoclavable Not Autoclavable
Sterility Non-sterile
Handle Material Wood
Product Type Applicator Stick
Tip Material Wood
Tip Type Single End