Learn More
Bochem™ Wood/Stainless Steel Diamant pencil
Product Code. 10709281
Click to view available options
Length (Metric):
150/135 mm
Unit Size:
Each
Product Code. 10709281
Brand: Bochem™ 12620
Only null left
Add to basket
Add to basket
BOchem™ Wood/Stainless Steel Diamant pencil can be used for cutting diamond.
  • Octahedron point
  • Wood/steel
  • Used in Cutting Diamond
Specifications
Product Type Diamant Pencil
Length (Metric) 150/135 mm
Material Wood, Steel
For Use With (Application) Used in Cutting Diamond
No. per Pack 1 Ea.