Learn More
Reichert Instruments™ Wolffheugel Counting Plate
Product Code. 12762585
Change view
Click to view available options
Quantity:
1Each
Unit Size:
Each
Product Code. Quantity Unit Size
12762585 1Each Each
1 options
Product Code. 12762585 Supplier Reichert Instruments™ Supplier No. 13333800
Estimated Shipment: 19-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Darkfield Quebec colony counter allows the user to quickly and accurately count bacteria colonies
The Darkfield Quebec colony counter allows the user to quickly and accurately count bacteria colonies.
It is available in two models for digital or manual counting. Both models provide even, glare-free illumination as light is spread uniformly over the entire culture plate.
Adjustable dish holder for centering both round dishes and square culture plates
  • Adjustable focusing rod
  • Lens rotates a full 360°
  • Built-in tilt leg
  • Optional 1.5x auxiliary lens fits over standard lens, increasing magnification to 3x
Specifications
Product Type Counting Plate
Quantity 1Each