missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Wogonin

Product Code. 16451398
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451398 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451398 Supplier MedChemExpress Supplier No. HYN0400

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Wogonin is a naturally occurring mono-flavonoid, can inhibit the activity of CDK8 and Wnt, and exhibits anti-inflammatory and anti-tumor effects.

Specifications

Content And Storage 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light)
Molecular Weight (g/mol) 284.26 g/mol
Shipping Condition Room temperature
For Use With (Application) Cancer-Kinase/protease
CAS 632-85-9
Product Type Natural Products
Purity 98.87%
Solubility DMSO : 62.5 mg/mL (219.87 mM∣Need ultrasonic)
Target CDK∣Wnt∣Autophagy∣Apoptosis
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.