missing translation for 'onlineSavingsMsg'
Learn More

wnvNS3 Protease Antibody (991926) [Allophycocyanin], Novus Biologicals™

Product Code. 30737502 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737502 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737502 Supplier Novus Biologicals Supplier No. FAB29073A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

wnvNS3 Protease Monoclonal antibody specifically detects wnvNS3 Protease in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen wnvNS3 Protease
Applications ELISA
Classification Monoclonal
Clone 991926
Conjugate APC
Formulation PBS
Host Species Mouse
Immunogen E. coli-derived Viral wnvNS3 Protease, sequences: rwnvNS2b 1-48, rwnvNS3 1-184
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 912267
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.