missing translation for 'onlineSavingsMsg'
Learn More

wnvNS3 Protease Antibody (345713) [Biotin], Novus Biologicals™

Product Code. 30733328 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30733328 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733328 Supplier Novus Biologicals Supplier No. FAB2907B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

wnvNS3 Protease Monoclonal antibody specifically detects wnvNS3 Protease in Virus samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen wnvNS3 Protease
Applications Western Blot
Classification Monoclonal
Clone 345713
Conjugate Biotin
Formulation PBS
Host Species Mouse
Immunogen E. coli-derived recombinant West Nile Virus NS3 Protease, expressed as a fusion protein with co-factor NS2L, Gly1506-Leu1689, Accession # ABD85079
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 912267
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.