missing translation for 'onlineSavingsMsg'
Learn More

wnvNS1 Protein Antibody (991551) [Janelia Fluor™ 669], Novus Biologicals™

Product Code. 30736449 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30736449 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30736449 Supplier Novus Biologicals Supplier No. FAB290722JF669

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

wnvNS1 Protein Monoclonal antibody specifically detects wnvNS1 Protein in Virus samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen wnvNS1 Protein
Applications ELISA
Classification Monoclonal
Clone 991551
Conjugate Janelia Fluor 669
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with recombinant Viral wnvNS1 Protein, Asp1-Ala352, Accession # NP_776015
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 912267
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.