Learn More
WNT8B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557723
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: PEFREVGAHL KEKYHAALKV DLLQGAGNSA AGRGAIADTF RSISTRELVH LEDSPDYCLE NKTLGLLGTE GRECLRRGRA LGRWER Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It encodes a protein which shows 95%, 86% and 71% amino acid identity to the mouse, zebrafish and Xenopus Wnt8B proteins, respectively. The expression patterns of the human and mouse genes appear identical and are restricted to the developing brain. The chromosomal location of this gene to 10q24 suggests it as a candidate gene for partial epilepsy.Specifications
WNT8B | |
Polyclonal | |
Unconjugated | |
WNT8B | |
protein Wnt-8b; wingless related MMTV integration site 8b; wingless-type MMTV integration site family member 8B; wingless-type MMTV integration site family, member 8B; WNT 8b; Wnt family member 8B; WNT8B; wnt-8b | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7479 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q93098 | |
WNT8B | |
Recombinant protein corresponding to Human WNT8B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only