Learn More
Invitrogen™ WNT8A Recombinant Rabbit Monoclonal Antibody (PSH0-39)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550826
Description
Positive control: HeLa cell lysate, HepG2 cell lysate, SW480 cell lysate, Jurkat cell lysate, Raji cell lysate, NIH/3T3 cell lysate, mouse brain tissue lysate, mouse embryo tissue lysate, rat brain tissue lysate, NIH/3T3, HeLa. Predicted band size: 38.8 kDa Subcellular Location: Secreted, extracellular space, extracellular matrix.
WNT8A, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.
Specifications
| WNT8A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q64527, Q9H1J5 | |
| Wnt8a | |
| Recombinant protein within human WNT8A aa 100-300/351. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| PSH0-39 | |
| Unconjugated | |
| Wnt8a | |
| etID309727.14; protein Wnt-8a; protein Wnt-8a ORF1; protein Wnt-8d; Stimulated by retinoic acid gene 11 protein; Stra11; wingless-related MMTV integration site 8A; wingless-related MMTV integration site 8D; wingless-type MMTV integration site family member 8A; wingless-type MMTV integration site family, member 8A; Wnt family member 8A; wnt8; wnt-8; wnt8.1; Wnt8a; Wnt-8A; Wnt8d; Wnt-8D; Wnt8-like protein 1; wu:fa20e02; wu:fe05d07 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 20890, 291678, 7478 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.