missing translation for 'onlineSavingsMsg'
Learn More

WNT5A/B Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 55184-1-AP-20UL

 View more versions of this product

Product Code. 16896665

  • £135.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Wnt5A is a secreted protein found in the extracellular region that belongs to the Wnt family. It is a proto-oncogene that functions in modulating cell fate and behavior during vertebrate development and has an important role in Wnt signaling pathway. It is a glycoprotein that plays an important role in cell communication and development. Wnt5A acts as a ligand for members of the frizzled family of seven transmembrane receptors. It is suspected that Wnt5A is a developmental protein. Wnt5A is a signaling molecule which affects the development of discrete regions of tissues and is likely to signal over only few cell diameters.
Specifications

Specifications

WNT5A/B
Polyclonal
Unconjugated
WNT5A
hWNT5A, Protein Wnt 5a, WNT5A, WNT5A B, WNT5A/B, WNT5B
Rabbit
Antigen Affinity Chromatography
RUO
22418, 22419, 7474, 81029
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P22725, P22726, P41221, Q9H1J7
WNT5A, WNT5B
Peptide
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.