Learn More
WNT4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534323
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Mouse, Rat, Hamster, Elephant, Panda, Bovine, Dog, Horse, Pig (100%) Opossum (86%).
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family, and is the first signaling molecule shown to influence the sex-determination cascade. It encodes a protein which shows 98% amino acid identity to the Wnt4 protein of mouse and rat. This gene and a nuclear receptor known to antagonize the testis-determining factor play a concerted role in both the control of female development and the prevention of testes formation. This gene and another two family members, WNT2 and WNT7B, may be associated with abnormal proliferation in breast tissue. Mutations in this gene can result in Rokitansky-Kuster-Hauser syndrome and in SERKAL syndrome.Specifications
WNT4 | |
Polyclonal | |
Unconjugated | |
Wnt4 | |
protein Wnt-4; SERKAL; signal molecule; UNQ426/PRO864; wingless-related MMTV integration site 4; wingless-type MMTV integration site family member 4; wingless-type MMTV integration site family, member 4; Wnt family member 4; WNT4; Wnt-4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100071678, 100327037, 100769003, 22417, 54361, 612367, 84426 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P22724, P56705, Q9QXQ5 | |
Wnt4 | |
Synthetic 14 amino acid peptide from internal region of human WNT4. | |
50 μg | |
Primary | |
Canine, Equine, Hamster, Human, Mouse, Non-human Primate, Porcine, Rat | |
Antibody | |
IgG |