missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNT11 Monoclonal Antibody (C4), Invitrogen™

Mouse Monoclonal Antibody
£178.00 - £408.00
Specifications
| Antigen | WNT11 |
|---|---|
| Clone | C4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17634006
|
Invitrogen™
MA543547 |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17624006
![]() |
Invitrogen™
MA543546 |
100 μL |
£408.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ligand for members of the frizzled family of seven transmembrane receptors. Probable developmental protein. May be a signaling molecule which affects the development of discrete regions of tissues. Is likely to signal over only few cell diameters.Specifications
| WNT11 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| HWNT11; Protein Wnt-11; wingless-related MMTV integration site 11; wingless-type MMTV integration site family, member 11; Wnt family member 11; Wnt11; Wnt-11 | |
| WNT11 | |
| IgG1 κ | |
| Protein A/G | |
| Antibody |
| C4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O96014 | |
| 7481 | |
| Ser204-Glu349 (Accession O96014), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| WNT11 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title