Learn More
Invitrogen™ WNT10B Monoclonal Antibody (5A7)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515557
Description
MA5-15557 targets WNT10B in indirect ELISA, IF, IHC, and WB applications and shows reactivity with Human samples. The MA5-15557 immunogen is purified recombinant fragment of human WNT10B expressed in E. Coli. MA5-15557 detects WNT10B which has a predicted molecular weight of approximately 43kDa.
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It may be involved in breast cancer, and its protein signaling is likely a molecular switch that governs adipogenesis. This protein is 96% identical to the mouse Wnt10b protein at the amino acid level. This gene is clustered with another family member, WNT1, in the chromosome 12q13 region.
Specifications
| WNT10B | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| O00744 | |
| WNT10B | |
| Purified recombinant fragment of human WNT10B expressed in E. Coli. | |
| RUO | |
| 7480 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 5A7 | |
| Unconjugated | |
| WNT10B | |
| Protein Wnt-10b; Protein Wnt-12; SHFM6; wingless related MMTV integration site 10b; wingless-type MMTV integration site family member 10B; wingless-type MMTV integration site family, member 10B; Wnt family member 10B; Wnt10; WNT10B; Wnt-10b; WNT-10B protein; WNT12; wnt-12 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.