missing translation for 'onlineSavingsMsg'
Learn More

PeproTech WNT1 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Product Code. 30054498
Change view
Click to view available options
Quantity:
2 x 500 μg
25 μg
50 μg
500 μg
Unit Size:
1mg
25µg
500µg
50µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30054498 500 μg 500µg
17885123 25 μg 25µg
17885433 50 μg 50µg
17834993 2 x 500 μg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30054498 Supplier PeproTech Supplier No. 500P250BT500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: ANSSGRWWGI VNVASSTNLL TDSKSLQLVL EPSLQLLSRK QRRLIRQNPG ILHSVSGGLQ SAVRECKWQF RNRRWNCPTA PGPHLFGKIV NRGCRETAFI FAITSAGVTH SVARSCSEGS IESCTCDYRR RGPGGPDWHW GGCSDNIDFG RLFGREFVDS GEKGRDLRFL MNLHNNEAGR TTVFSEMRQE CKCHGMSGSC TVRTCWMRLP TLRAVGDVLR DRFDGASRVL YGNRGSNRAS RAELLRLEPE DPAHKPPSPH DLVYFEKSPN FCTYSGRLGT AGTAGRACNS SSPALDGCEL LCCGRGHRTR TQRVTERCNC TFHWCCHVSC RNCTHTRVLH ECL. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human Wnt-1. Anti-Human Wnt-1-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hWNT-1 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human WNT-1 (500-P250) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hWNT-1. Western Blot: To detect hWNT-1 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hWNT-1 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P250BT-1MG will be provided as 2 x 500 μg

Wnt proteins constitute a family of secreted glyocoproteins that activate signal transduction pathways to mediate tissue homeostasis, cell fate, cell proliferation, and self-renewal. The canonical signaling pathway is initiated upon Wnt binding to a Frizzled family receptor and an LRP5/LRP6 coreceptor, which leads to activation of the beta-catenin cascade. Non-canonical Wnt signals are activated by binding to a Frizzled family receptor and ROR2 coreceptor. Extensive research has shown that deregulation of these pathways or its components can lead to cancer. Human Wnt-1 is encoded as a 370 amino acid precursor that, upon secretion, is cleaved into a 343 amino acid (42 kDa) mature form. At the amino acid level, mature human Wnt-1 is approximately 99% identical to mature mouse Wnt-1. Depending on the tissue, Wnt-1 can activate the canonical and non-canonical signaling pathways. Studies have demonstrated its involvement in a variety of functions, including embryonic development and carcinogenesis. Wnt-1 signaling can also induce the phosphorylation and subsequent degradation of c-Myb.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNT1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene WNT1
Gene Accession No. P04628
Gene Alias BMND16; INT1; Int-1; murine mammary tumor virus integration site; OI15; Proto-oncogene Int-1; proto-oncogene Int-1 homolog; proto-oncogene protein Wnt-1; proto-oncogene Wnt-1; sw; swaying; wingless-related MMTV integration site 1; Wingless-type MMTV integration site 1 homolog; Wingless-type MMTV integration site 1, homolog; wingless-type MMTV integration site family member 1; wingless-type MMTV integration site family, member 1; wingless-type MMTV integration site family, member 1 (oncogene INT1); Wnt family member 1; Wnt1; Wnt-1
Gene Symbols WNT1
Host Species Rabbit
Immunogen E.coli-derived, 38.4 kDa Recombinant Human Wnt-1.
Purification Method Antigen affinity chromatography
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7471
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.