missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Wnt-C59
GREENER_CHOICE

Product Code. p-5546108
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18702459

Brand: Tocris Bioscience™ 5148/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent PORCN inhibitor

Wnt-C59 is a highly potent inhibitor of MBOAT (membrane-bound O-acyltranferase) family member Porcupine (PORCN) (IC50 = 74 pM) that mediates WNT palmitoylation and secretion. Wnt-C59 potently inhibits the processing of both canonical (1, 2, 3a, 6, 7b, 8a, 9a, 9b, 10) and non-canonical (4, 5a, 11, 16) Wnt subtypes. Wnt-C59 blocks progression of mammary tumors in MMTV-WNT1 transgenic mice and downregulates Wnt/β-catenin target genes. Wnt-C59 treated tumors show a decrease in β-catenincatenin, CyclinD1 and c-Myc. Wnt-C59 induces cardiomyocyte differentiation from human iPSCs following culture with CHIR 99021 (Cat. No. 4423). Wnt-C59 efficiently induces neural differentiation of CTIP2+/COUP-TF1- cells from PSCs in culture. When grafted into the cortex of adult mice, develop abundant axonal fiber extension toward the spinal cord. Cell permeable and orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type Wnt-C59
Purity >99%
Molecular Weight (g/mol) 379.45
Inhibitors Wnt-C59

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.