missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Wnt-C59

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Wnt-C59 is a highly potent inhibitor of MBOAT (membrane-bound O-acyltranferase) family member Porcupine (PORCN) (IC50 = 74 pM) that mediates WNT palmitoylation and secretion. Wnt-C59 potently inhibits the processing of both canonical (1, 2, 3a, 6, 7b, 8a, 9a, 9b, 10) and non-canonical (4, 5a, 11, 16) Wnt subtypes. Wnt-C59 blocks progression of mammary tumors in MMTV-WNT1 transgenic mice and downregulates Wnt/β-catenin target genes. Wnt-C59 treated tumors show a decrease in β-catenincatenin, CyclinD1 and c-Myc. Wnt-C59 induces cardiomyocyte differentiation from human iPSCs following culture with CHIR 99021 (Cat. No. 4423). Wnt-C59 efficiently induces neural differentiation of CTIP2+/COUP-TF1- cells from PSCs in culture. When grafted into the cortex of adult mice, develop abundant axonal fiber extension toward the spinal cord. Cell permeable and orally bioavailable.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | Wnt-C59 |
| Purity | >99% |
| Molecular Weight (g/mol) | 379.45 |
| Inhibitors | Wnt-C59 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction