missing translation for 'onlineSavingsMsg'
Learn More

Wnt-9b Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NBP1-51961

Product Code. 18260695

  • £407.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Wnt-9b Polyclonal specifically detects Wnt-9b in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Wnt-9b
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Protein Wnt-14b, wingless-type MMTV integration site family, member 15, wingless-type MMTV integration site family, member 9B, WNT14Bprotein Wnt-9b, WNT15Protein Wnt-15
Goat
Affinity Purified
RUO
Primary
Cross reacts with Human. Expected from sequence similarity: Mouse and Rat.
Store at -20C. Avoid freeze-thaw cycles.
Western Blot, ELISA
0.5 mg/mL
Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:8000
NP_003387.1
WNT9B
Peptide with sequence C-KRGNKDLRARADA corresponding to internal region according to NP_003387.1.
0.1 mg
Signal Transduction, Wnt Signaling Pathway
7484
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only