missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-8a Antibody (PSH0-39), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33103
This item is not returnable.
View return policy
Description
Wnt-8a Monoclonal antibody specifically detects Wnt-8a in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Wnt-8a | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50 | |
| protein Wnt-8a, Protein Wnt-8d, wingless-type MMTV integration site family, member 8A, WNT8DWNT8d | |
| Recombinant protein within human Wnt-8a aa 100-300/ 351. (Uniprot: Q9H1J5) | |
| 100 μL | |
| Signal Transduction, Wnt Signaling Pathway | |
| 7478 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| PSH0-39 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction