missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-8a Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-14523
This item is not returnable.
View return policy
Description
Wnt-8a Polyclonal antibody specifically detects Wnt-8a in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Wnt-8a | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| protein Wnt-8a, Protein Wnt-8d, wingless-type MMTV integration site family, member 8A, WNT8DWNT8d | |
| Synthetic 18 amino acid peptide from internal region of human Wnt-8a. | |
| 50 μg | |
| Signal Transduction, Wnt Signaling Pathway | |
| 7478 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction