missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-7b Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 4 publications
Brand: Novus Biologicals NBP1-59564
This item is not returnable.
View return policy
Description
Wnt-7b Polyclonal specifically detects Wnt-7b in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| Wnt-7b | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| protein Wnt-7b, wingless-type MMTV integration site family, member 7B | |
| Rabbit | |
| 36 kDa | |
| 100 μL | |
| Signal Transduction, Wnt Signaling Pathway | |
| 7477 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 4-8 ug/ml, Immunohistochemistry-Frozen | |
| P56706 | |
| WNT7B | |
| Synthetic peptides corresponding to WNT7B(wingless-type MMTV integration site family, member 7B) The peptide sequence was selected from the middle region of WNT7B (NP_478679) Peptide sequence WTTLPKFREVGHLLKEKYNAAVQVEVVRASRLRQPTFLRIKQLRSYQKPM. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Expected identity based on immunogen sequence: Xenopus: 78%; Chicken: 78%. | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction