missing translation for 'onlineSavingsMsg'
Learn More

Wnt-7b Antibody, Novus Biologicals™

Product Code. 18321139 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18321139 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18321139 Supplier Novus Biologicals Supplier No. H00007477B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

Wnt-7b Polyclonal antibody specifically detects Wnt-7b in Human, Mouse samples. It is validated for Western Blot, Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen Wnt-7b
Applications Western Blot, Functional Assay
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Functional
Formulation PBS (pH 7.4)
Gene Accession No. NP_478679.1
Gene Alias protein Wnt-7b, wingless-type MMTV integration site family, member 7B
Host Species Mouse
Immunogen WNT7B (NP_478679.1, 1 a.a. - 349 a.a.) full-length human protein. MHRNFRKWIFYVFLCFGVLYVKLGALSSVVALGANIICNKIPGLAPRQRAICQSRPDAIIVIGEGAQMGINECQYQFRFGRWNCSALGEKTVFGQELRVGSREAAFTYAITAAGVAHAVTAACSQGNLSNCGCDREKQGYYNQAEGWKWGGCSADVRYGIDFSRRFVDAREIKKNARRLMNLHNNEAGRKVLEDRMQLECKCHGVSGSCTTKTCWTTLPKFREVGHLLKEKYNAAVQVEVVRASRLRQPTFLRIKQLRSYQKPMETDLVYIEKSPNYCEEDAATGSVGTQGRLCNRTSPGADGCDTMCCGRGYNTHQYTKVWQCNCKFHWCCFVKCNTCSERTEVFTCK
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7477
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.