missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-7b Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93321-0.1ml
This item is not returnable.
View return policy
Description
Wnt-7b Polyclonal antibody specifically detects Wnt-7b in Human samples. It is validated for Western Blot
Specifications
| Wnt-7b | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| protein Wnt-7b, wingless-type MMTV integration site family, member 7B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 120-349 of human WNT7B (NP_478679.1). TAACSQGNLSNCGCDREKQGYYNQAEGWKWGGCSADVRYGIDFSRRFVDAREIKKNARRLMNLHNNEAGRKVLEDRMQLECKCHGVSGSCTTKTCWTTLPKFREVGHLLKEKYNAAVQVEVVRASRLRQPTFLRIKQLRSYQKPMETDLVYIEKSPNYCEEDAATGSVGTQGRLCNRTSPGADGCDTMCCGRGYNTHQYTKVWQCNCKFHWCCFVKCNTCSERTEVFTCK | |
| 0.1 mL | |
| Signal Transduction, Wnt Signaling Pathway | |
| 7477 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction