missing translation for 'onlineSavingsMsg'
Learn More

Wnt-7a Antibody (946744) [DyLight 405], Novus Biologicals™

Product Code. 30731871 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30731871 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731871 Supplier Novus Biologicals Supplier No. FAB30081E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Wnt-7a Monoclonal antibody specifically detects Wnt-7a in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Wnt-7a
Applications Immunocytochemistry
Classification Monoclonal
Clone 946744
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias protein Wnt-7a, proto-oncogene Wnt7a protein, wingless-type MMTV integration site family, member 7A
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Wnt-7a, Met1-Lys349, Accession # O00755
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7476
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.