missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-5b Antibody (SR1784), Novus Biologicals™
Rabbit Monoclonal Antibody
£257.00 - £341.00
Specifications
| Antigen | Wnt-5b |
|---|---|
| Clone | SR1784 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50 |
| Applications | Western Blot, Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620755
|
Novus Biologicals
NBP3-22311-100ul |
100 μg |
£341.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669364
|
Novus Biologicals
NBP3-22311-50ul |
50 μg |
£257.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Wnt-5b Monoclonal antibody specifically detects Wnt-5b in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
| Wnt-5b | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC2648, protein Wnt-5b, wingless-type MMTV integration site family, member 5B, WNT-5B protein | |
| A synthesized peptide derived from human Wnt-5b (Uniprot #: Q9H1J7) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1784 | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 81029 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title