missing translation for 'onlineSavingsMsg'
Learn More

Wnt-5b Antibody (325004) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30735490 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735490 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735490 Supplier Novus Biologicals Supplier No. FAB3006JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Wnt-5b Monoclonal antibody specifically detects Wnt-5b in Mouse samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Wnt-5b
Applications Immunocytochemistry
Classification Monoclonal
Clone 325004
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias MGC2648, protein Wnt-5b, wingless-type MMTV integration site family, member 5B, WNT-5B protein
Host Species Rat
Immunogen E. coli-derived recombinant mouse Wnt-5b, Ser55-Ser95 and Lys246-Glu315 connected by a Gly-Ser linker, Accession # NP_033551
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 81029
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.