missing translation for 'onlineSavingsMsg'
Learn More

Wnt-5a Rabbit anti-Human, Clone: 3P2E8, Novus Biologicals™

Product Code. 18368416 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18368416 20 μg 20µL
18356856 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18368416 Supplier Novus Biologicals Supplier No. NBP31568420UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Wnt-5a Monoclonal antibody specifically detects Wnt-5a in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Wnt-5a
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3P2E8
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias hWNT5A, protein Wnt-5a, wingless-type MMTV integration site family, member 5A, WNT-5A protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 281-380 of human Wnt-5a (P41221).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer Stem Cells, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7474
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.