missing translation for 'onlineSavingsMsg'
Learn More

Wnt-5a Antibody (S08-4A3), Novus Biologicals™

Rabbit Monoclonal Antibody

£328.00 - £500.00

Specifications

Antigen Wnt-5a
Clone S08-4A3
Concentration 0.3 mg/mL
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18603084
View Documents
Novus Biologicals
NBP3-20061-100ul
100 μg
£500.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18683785
View Documents
Novus Biologicals
NBP3-20061-50ul
50 μg
£328.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Wnt-5a Monoclonal antibody specifically detects Wnt-5a in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), RNA Inhibition, Dot Blot, Sandwich ELISA
Specifications

Specifications

Wnt-5a
0.3 mg/mL
Monoclonal
Purified
RUO
Human
hWNT5A, protein Wnt-5a, wingless-type MMTV integration site family, member 5A, WNT-5A protein
A synthetic peptide of human Wnt-5a (Uniprot # P41221)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
S08-4A3
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Rabbit
Cancer Stem Cells, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
7474
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.