Learn More
Wnt-5a Antibody (CL13683) [Janelia Fluor™ 646], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24567JF646
Product Code. 30297644
  • £558.00 / 0.10mL
Description
Wnt-5a Monoclonal antibody specifically detects Wnt-5a in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Wnt-5a
Monoclonal
Janelia Fluor 646
hWNT5A, protein Wnt-5a, wingless-type MMTV integration site family, member 5A, WNT-5A protein
This antibody was developed using a synthetic peptide derived from P41221, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cancer Stem Cells, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
7474
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL13683
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified