missing translation for 'onlineSavingsMsg'
Learn More

Wnt-16b Antibody (948309) [CoraFluor™ 1], Novus Biologicals™

Product Code. 30645038 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30645038 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30645038 Supplier Novus Biologicals Supplier No. FAB7790CL1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Wnt-16b Monoclonal antibody specifically detects Wnt-16b in Human samples. It is validated for Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Wnt-16b
Applications Immunocytochemistry
Classification Monoclonal
Clone 948309
Conjugate CoraFluor 1
Formulation PBS
Gene Alias AHCP
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Wnt-16, Asn30-Lys365, Accession # Q9UBV4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51384
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.