missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31406
This item is not returnable.
View return policy
Description
Wnt-11 Polyclonal antibody specifically detects Wnt-11 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| Wnt-11 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0) and 20% Glycerol with 0.025% Proclin 300 | |
| HWNT11, MGC141946, MGC141948, protein Wnt-11, wingless-type MMTV integration site family, member 11 | |
| Rabbit | |
| 39 kDa | |
| 100 μL | |
| Wnt Signaling Pathway | |
| 7481 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| O96014 | |
| WNT11 | |
| Recombinant protein encompassing a sequence within the center region of human WNT11. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Predicted cross-reactivity based on sequence identity: Chicken (85%), Xenopus laevis (81%), Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction