missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wnt-10b Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77325MFV450
This item is not returnable.
View return policy
Description
Wnt-10b Polyclonal antibody specifically detects Wnt-10b in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Wnt-10b | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| protein Wnt-10b, Protein Wnt-12, SHFM6WNT-10B protein, wingless-type MMTV integration site family, member 10B, WNT12, WNT-12 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the center of human Wnt10b. The immunogen is located within amino acids 130 - 180 of Wnt10b. Amino Acid Squence: CGWKGSGEQDRLRAK | |
| 0.1 mL | |
| Wnt Signaling Pathway | |
| 7480 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction