missing translation for 'onlineSavingsMsg'
Learn More

Wnt-10b Antibody, DyLight 680, Novus Biologicals™

Product Code. 18429957 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429957 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429957 Supplier Novus Biologicals Supplier No. NBP177325FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Wnt-10b Polyclonal antibody specifically detects Wnt-10b in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Wnt-10b
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias protein Wnt-10b, Protein Wnt-12, SHFM6WNT-10B protein, wingless-type MMTV integration site family, member 10B, WNT12, WNT-12
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the center of human Wnt10b. The immunogen is located within amino acids 130 - 180 of Wnt10b. Amino Acid Squence: CGWKGSGEQDRLRAK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7480
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.