missing translation for 'onlineSavingsMsg'
Learn More

WNK463, MedChemExpress

Product Code. 16450679
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16450679 1 mg 1mg
16443108 10 mg 10mg
16409666 50 mg 50mg
16404406 100 mg 100mg
16485837 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16450679 Supplier Medchem Express Supplier No. HY100626

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress WNK463 is an orally bioavailable pan-With-No-Lysine (K) (WNK)-kinase inhibitor with IC50s of 5 nM, 1 nM, 6 nM, and 9 nM for WNK1, WNK2, WNK3, and WNK4, respectively.

WNK463 is an orally bioavailable pan-With-No-Lysine (K) (WNK)-kinase inhibitor with IC50s of 5 nM, 1 nM, 6 nM, and 9 nM for WNK1, WNK2, WNK3, and WNK4, respectively.

Specifications

Chemical Name or Material WNK463
CAS 2012607-27-9
Color White
Purity Grade Notes Research
Molecular Formula C21H24F3N7O2
Quantity 1 mg
Solubility Information DMSO : ≥ 30 mg/mL (64.73 mM)
SMILES FC(F)(F)C1=NN=C(O1)C2=CN=C(N3CCC(N4C=NC=C4C(NC(C)(C)C)=O)CC3)C=C2
Molecular Weight (g/mol) 463.46
Formula Weight 463.46
Percent Purity 99.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.