missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WNK4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22326-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the WNK family of serine-threonine protein kinases. The kinase is part of the tight junction complex in kidney cells, and regulates the balance between NaCl reabsorption and K(+) secretion. The kinase regulates the activities of several types of ion channels, cotransporters, and exchangers involved in electrolyte flux in epithelial cells. Mutations in this gene result in pseudohypoaldosteronism type IIB.Specifications
| WNK4 | |
| Polyclonal | |
| Unconjugated | |
| WNK4 | |
| PHA2B, PRKWNK4, WNK4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 65266, 69847 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q80UE6, Q96J92 | |
| Wnk4 | |
| WNK4 Fusion Protein Ag17773 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction