missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WNK3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA590519
This item is not returnable.
View return policy
Description
Immunogen sequence: AGCLEERRDS QCKSMGNVFP QPQNTTLPLA PAQQTGAECE ETEVDQHVRQ QLLQRKPQQH CSSVTGDNLS EAGAASVIHS DTSSQPSVAY SSNQTMGSQM V; Positive Samples: Mouse brain; Cellular Location: Cytoplasm.
WNK3 is a serine/threonine kinase involved in signal transduction and cellular communication. Members of the 'with no lysine' kinase family, such as WNK3, are serine-threonine protein kinases that lack the almost invariant catalytic lysine in subdomain II, which is important for binding ATP in the catalytic site. Instead, these kinases have a conserved lysine in subdomain I that is thought to provide this function.
Specifications
| WNK3 | |
| Polyclonal | |
| Unconjugated | |
| WNK3 | |
| KIAA1566; PRKWNK 3; PRKWNK3; Protein kinase lysine-deficient 3; protein kinase with no lysine 3; RGD1563131; serine/threonine-protein kinase WNK3; WNK 3; WNK lysine deficient protein kinase 3; WNK lysine deficient protein kinase 3, pseudogene; WNK3; Wnk3-ps | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 279561, 317420, 65267 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.29 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q80XP9, Q9BYP7 | |
| WNK3 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction