missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WNK2 Antibody (2A2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00065268-M02
This item is not returnable.
View return policy
Description
WNK2 Monoclonal antibody specifically detects WNK2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| WNK2 | |
| Monoclonal | |
| Unconjugated | |
| NP_006639 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 2A2 | |
| In 1x PBS, pH 7.4 | |
| EC 2.7.11, EC 2.7.11.1, PRK, PRKWNK2, Protein kinase with no lysine 2, SDCCAG43, serine/threonine-protein kinase WNK2, Serologically defined colon cancer antigen 43lysine deficient 2, WNK lysine deficient protein kinase 2 | |
| WNK2 (NP_006639, 2118 a.a. ∽ 2217 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KGTFTDDLHKLVDEWTSKTVGAAQLKPTLNQLKQTQKLQDMEAQAGWAAPGEARAMTAPRAGVGMPRLPPAPGPLSTTVIPGAAPTLSVPTPDPESEKPD | |
| 0.1 mg | |
| Protein Kinase | |
| 65268 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction