missing translation for 'onlineSavingsMsg'
Learn More

WNK1 Rabbit anti-Mouse, Clone: 6C5S6, Novus Biologicals™

Product Code. 18329696 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18329696 100 μg 100µL
18336215 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18329696 Supplier Novus Biologicals Supplier No. NBP316360100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

WNK1 Monoclonal antibody specifically detects WNK1 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen WNK1
Applications Western Blot
Classification Monoclonal
Clone 6C5S6
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 2.7.11, EC 2.7.11.1, Erythrocyte 65 kDa protein, HSAN2hereditary sensory neuropathy, type II, HSN2MGC163339, hWNK1, KDPMGC163341, KIAA0344, Kinase deficient protein, p65, PHA2C, PRKWNK1PSK, prostate-derived sterile 20-like kinase, Protein kinase lysine-deficient 1, Protein kinase with no lysine 1, protein kinase, lysine deficient 1, serine/threonine-protein kinase WNK1, WNK lysine deficient protein kinase 1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human WNK1 (Q9H4A3). MSGGAAEKQSSTPGSLFLSPPAPAPKNGSSSDSSVGEKLGAAAADAVTGRTEEYRRRRHTMDKDSRGAAATTTTTEHRFFRRSVICDSNATALELPGLPL
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 65125
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.