missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WNK1 Antibody (26G7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26251-50ul
This item is not returnable.
View return policy
Description
WNK1 Monoclonal antibody specifically detects WNK1 in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| WNK1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 26G7 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| EC 2.7.11, EC 2.7.11.1, Erythrocyte 65 kDa protein, HSAN2hereditary sensory neuropathy, type II, HSN2MGC163339, hWNK1, KDPMGC163341, KIAA0344, Kinase deficient protein, p65, PHA2C, PRKWNK1PSK, prostate-derived sterile 20-like kinase, Protein kinase lysine-deficient 1, Protein kinase with no lysine 1, protein kinase, lysine deficient 1, serine/threonine-protein kinase WNK1, WNK lysine deficient protein kinase 1 | |
| A synthesized peptide derived from Human WNK1 [UniProt Q9H4A3] | |
| 50 μL | |
| Phospho Specific, Protein Kinase, Signal Transduction, Transcription Factors and Regulators | |
| 65125 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction