missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ WM 1119

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
WM 1119 is a high affinity and competitive lysine acetyltransferase 6A (KAT6A/MOZ) inhibitor (Kd = 2 nM), which displays >250-fold affinity for KAT6A compared to KAT5 and KAT7. WM 1119 induces cell-cycle exit and cellular senescence, and inhibits growth of B-cell lymphoma cells in vitro (IC50 = 0.25 μM). WM 1119 also reduces H3K9ac in tumor cells, and decreases tumor burden and spleen weight in a mouse lymphoma model.
Specifications
Specifications
| Chemical Name or Material | 2-Fluoro-N'-(3-fluoro-5-(pyridin-2-yl)benzoyl)benzenesulfonohydrazide |
| CAS | 2055397-28-7 |
| Quantity | 10 mg |
| Molecular Formula | C18H13F2N3O3S |
| Formula Weight | 389.38 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction