missing translation for 'onlineSavingsMsg'
Learn More

WM-1119, MedChemExpress

Product Code. 16442836
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16442836

Brand: Medchem Express HY102058

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WM-1119 is a highly potent and selective KAT6A inhibitor, with an IC50 of 0.25 μM for KAT6A in lymphoma cells, the binding KD values of WM-1119 with KAT6A, KAT5 and KAT7 are 2 nM, 2.2 μM, 0.5 μM , respectively.

WM-1119 is a highly potent and selective KAT6A inhibitor, with an IC50 of 0.25 μM for KAT6A in lymphoma cells, the binding KD values of WM-1119 with KAT6A, KAT5 and KAT7 are 2 nM, 2.2 μM, 0.5 μM , respectively.

Specifications

Chemical Name or Material WM-1119
CAS 2055397-28-7
Color Light Nude
Purity Grade Notes Research
Molecular Formula C18H13F2N3O3S
Quantity 25 mg
Solubility Information DMSO : 125 mg/mL (321.02 mM; Need ultrasonic)
SMILES O=C(NNS(=O)(C1=CC=CC=C1F)=O)C2=CC(C3=NC=CC=C3)=CC(F)=C2
Molecular Weight (g/mol) 389.38
Formula Weight 389.38
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.