Learn More
Thermo Scientific™ WKITS™ Water QC Kit 1003
Description
The Thermo Scientific™ WKIT Water QC Kit 1003 is used for water/wastewater quality control testing; microorganisms are supplied in Quanti-Cult format.
- WaterBugs™ deliver <50 CFUs in a single inoculum to mimic water samples
Ordering Alerts:
Reagents are sufficient for one test/reconstituted vial.
Includes:
Contents: 10 preserved organism vials (5 per organism); 10 rehydration vials. Escherichia coli strain C1, Pseudomonas aeruginosa ATCC™ 27853™†
Specifications
Specifications
| Description | Water QC Kit 1003 |
| Product Type | Quality Control Kit |
| Quantity | 2/Pk. |
The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.