missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Withanolide A, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | Withanolide A |
| CAS | 32911-62-9 |
| Molecular Formula | C28 H38 O6 |
| Quantity | 2.5 mg |
| Synonym | (5α,6α,7α,22R)-6,7-epoxy-5,20,22-trihydroxy-1-oxo-ergosta-2,24-dien-26-oic Acid Δ-Lactone, (22R)-6α,7α-epoxy-5,20,22-trihydroxy-1-oxo-5α-Ergosta-2,24-dien-26-oic Acid Δ-Lactone, 6,7-Epoxy-1H-cyclopenta[a]phenanthrene Ergosta-2,24-dien-26-oic Acid Deriv., 5,20α-Dihydroxy-6α,7α-epoxy-1-oxowitha-2,24-dienolide, 5α,20αF(R)-Dihydroxy-6α,7α-epoxy-1-oxowitha-2,24-dienolide, Withaniol |
| InChI Formula | InChI=1S/C28H38O6/c1-14-13-20(33-24(30)15(14)2)27(5,31)18-9-8-16-21-17(10-12-25(16,18)3)26(4)19(29)7-6-11-28(26,32)23-22(21)34-23/h6-7,16-18,20-23,31-32H,8-13H2,1-5H3/t16-,17-,18-,20+,21-,22-,23-,25-,26-,27+,28-/m0/s1 |
| SMILES | CC1=C(C)C(=O)O[C@H](C1)[C@](C)(O)[C@H]2CC[C@H]3[C@@H]4[C@@H]5O[C@@H]5[C@@]6(O)CC=CC(=O)[C@]6(C)[C@H]4CC[C@]23C |
| Molecular Weight (g/mol) | 470.6 |
| Formula Weight | 470.2668 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction