missing translation for 'onlineSavingsMsg'
Learn More

WISP3 Antibody, Novus Biologicals™

Product Code. 18336748 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18336748 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18336748 Supplier Novus Biologicals Supplier No. H00008838B01P50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

WISP3 Polyclonal antibody specifically detects WISP3 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen WISP3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS (pH 7.4)
Gene Accession No. NP_937882.1
Gene Alias CCN family member 6, CCN6MGC125988, LIBC, MGC125987, MGC125989, PPAC, PPD, WISP-3, WNT1 inducible signaling pathway protein 3, WNT1-inducible-signaling pathway protein 3
Host Species Mouse
Immunogen WISP3 (NP_937882.1, 1 a.a. - 372 a.a.) full-length human protein. MNKRRLLYPSGWLHGPSDMQGLLFSTLLLAGLAQFCCRVQGTGPLDTTPEGRPGEVSDAPQRKQFCHWPCKCPQQKPRCPPGVSLVRDGCGCCKICAKQPGEICNEADLCDPHKGLYCDYSVDRPRYETGVCAYLVAVGCEFNQVHYHNGQVFQPNPLFSCLCVSGAIGCTPLFIPKLAGSHCSGAKGGKKSDQSNCSLEPLLQQLSTSYKTMPAYRNLPLIWKKKCLVQATKWTPCSRTCGMGISNRVTNENSNCEMRKEKRLCYIQPCDSNILKTIKIPKGKTCQPTFQLSKAEKFVFSGCSSTQSYKPTFCGICLDKRCCIPNKSKMITIQFDCPNEGSFKWKMLWITSCVCQRNCREPGDIFSELKIL
Purification Method Immunogen affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 8838
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.