missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WISP2, Mouse anti-Human, Clone: 3D10, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00008839-M09.100ug
This item is not returnable.
View return policy
Description
Sequence: QLCPTPCTCPWPPPRCPLGVPLVLDGCGCCRVCARRLGEPCDQLHVCDASQGLVCQPGAGPGGRGALCLLAEDDCSCEVNGRLYREGETFQPHCSIRCRCEDGGFTCVPLCSEDVRLPSWDCPHPRRVEVLGKCCPEWVCGQGGGLGTQPLPAQGPQFSGLVSSLPPGVPCPEWSTAWGPCSTTCGLGMATRVSNQNRFCRLETQRRLCLSRPCPPSRGRSPQNSAFSpecifications
| WISP2 | |
| Monoclonal | |
| Unconjugated | |
| WNT1 inducible signaling pathway protein 2 | |
| CCN5/CT58/CTGF-L | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunoprecipitation, Western Blot | |
| 3D10 | |
| In 1x PBS, pH 7.4 | |
| BC017782 | |
| WISP2 | |
| WISP2 (AAH17782.1, 24 a.a. ∼ 250 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 8839 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction