missing translation for 'onlineSavingsMsg'
Learn More

WISP2 Antibody (1E11), Novus Biologicals™

Product Code. 18398138 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18398138 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18398138 Supplier Novus Biologicals Supplier No. H00008839M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

WISP2 Monoclonal antibody specifically detects WISP2 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen WISP2
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1E11
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH17782
Gene Alias Connective tissue growth factor-like protein, Connective tissue growth factor-related protein 58, CT58CTGF-Lwnt-1 signaling pathway protein 2, CTGFL, WISP-2, WNT1 inducible signaling pathway protein 2, WNT1-inducible-signaling pathway protein 2
Host Species Mouse
Immunogen WISP2 (AAH17782.1, 24 a.a. ~ 250 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. QLCPTPCTCPWPPPRCPLGVPLVLDGCGCCRVCARRLGEPCDQLHVCDASQGLVCQPGAGPGGRGALCLLAEDDCSCEVNGRLYREGETFQPHCSIRCRCEDGGFTCVPLCSEDVRLPSWDCPHPRRVEVLGKCCPEWVCGQGGGLGTQPLPAQGPQFSGLVSSLPPGVPCPEWSTAWGPCSTTCGLGMATRVSNQNRFCRLETQRRLCLSRPCPPSRGRSPQNSAF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Cardiovascular Biology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 8839
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.