missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WISP1 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA547183
This item is not returnable.
View return policy
Description
In Western blots, approximately 10% cross-reactivity with recombinant human (rh) WISP-1 is observed and less than 1% cross-reactivity with rhCTGF and recombinant mouse NOV is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the WNT1 inducible signaling pathway (WISP) protein subfamily, which belongs to the connective tissue growth factor (CTGF) family. WNT1 is a member of a family of cysteine-rich, glycosylated signaling proteins that mediate diverse developmental processes. The CTGF family members are characterized by four conserved cysteine-rich domains: insulin-like growth factor-binding domain, von Willebrand factor type C module, thrombospondin domain and C-terminal cystine knot-like domain. This gene may be downstream in the WNT1 signaling pathway that is relevant to malignant transformation. It is expressed at a high level in fibroblast cells, and overexpressed in colon tumors. The encoded protein binds to decorin and biglycan, two members of a family of small leucine-rich proteoglycans present in the extracellular matrix of connective tissue, and possibly prevents the inhibitory activity of decorin and biglycan in tumor cell proliferation. It also attenuates p53-mediated apoptosis in response to DNA damage through activation of the Akt kinase. It is 83% identical to the mouse protein at the amino acid level. Alternative splicing of this gene generates 2 transcript variants.
Specifications
| WISP1 | |
| Polyclonal | |
| Unconjugated | |
| CCN4 | |
| AW146261; CCN family member 4; Ccn4; CTC-458A3.8; Elm1; ELM-1; WISP1; WISP-1; WISP1c; WISP1i; WISP1tc; WNT1 induced secreted protein 1; WNT1 inducible signaling pathway protein 1; Wnt-1 inducible signaling pathway protein 1; Wnt-1-induced secreted protein; WNT1-inducible-signaling pathway protein 1 | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 22402 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Frozen), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O54775 | |
| CCN4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse WISP-1/CCN4 Thr23-Asn367. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction