missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WISP1 Human ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EHWISP1X5

Additional Details : Weight : 0.33820kg

 View more versions of this product

Product Code. 17195599

  • £2320.00 / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

The Human WISP-1 ELISA quantitates Hu WISP-1 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu WISP-1. Principle of the method The Human WISP-1 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes a member of the WNT1 inducible signaling pathway (WISP) protein subfamily, which belongs to the connective tissue growth factor (CTGF) family. WNT1 is a member of a family of cysteine-rich, glycosylated signaling proteins that mediate diverse developmental processes. The CTGF family members are characterized by four conserved cysteine-rich domains: insulin-like growth factor-binding domain, von Willebrand factor type C module, thrombospondin domain and C-terminal cystine knot-like domain. This gene may be downstream in the WNT1 signaling pathway that is relevant to malignant transformation. It is expressed at a high level in fibroblast cells, and overexpressed in colon tumors. The encoded protein binds to decorin and biglycan, two members of a family of small leucine-rich proteoglycans present in the extracellular matrix of connective tissue, and possibly prevents the inhibitory activity of decorin and biglycan in tumor cell proliferation. It also attenuates p53-mediated apoptosis in response to DNA damage through activation of the Akt kinase. It is 83% identical to the mouse protein at the amino acid level. Alternative splicing of this gene generates 2 transcript variants.
Specifications

Specifications

O95388
50 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
5 x 96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
8840
<12%
Stop Solution, Standard, Wash Buffer, SAV-HRP, Adhesive Plate Covers, Biotinylated Detection Antibody, Pre-coated 96 well plates, Chromogen, Assay Diluent Concentrate
5 x 96 Tests
Development
2°C to 8°C
Human
4 hr. 45 min.
61.44 pg/mL to 15,000 pg/mL
50 pg/mL to 15,000 pg/mL
Biotin
Other Proteins
In this sandwich ELISA format, no cross-reactivity was observed with rhBiglycan, rhDecorin, rhNOV, or rhWISP-3.
Human
Colorimetric Microplate Reader
CCN4c,WISP1i,WISP1tc,WISP1
<10%
HRP
RUO
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L
CCN4c, WISP1i, WISP1tc, WISP1
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers