missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
WISP-1/CCN4 Polyclonal specifically detects WISP-1/CCN4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | WISP-1/CCN4 |
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml |
| Formulation | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Gene Alias | CCN family member 4, CCN4WISP1tc, WISP-1, WISP1c, WISP1i, WNT1 induced secreted protein 1, Wnt-1 inducible signaling pathway protein 1, WNT1 inducible signaling pathway protein 1, wnt-1 signaling pathway protein 1, Wnt-1-induced secreted protein, WNT1-inducible-signaling pathway protein 1 |
| Gene Symbols | CCN4 |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:QVVGVGCVLDGVRYNNGQSFQPNCKYNCTCIDGAVGCTPLCLRVRPPRLWCPHPRRVSIPGHCCEQWVCEDDAKRPRKTAPRDTGAF |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?