missing translation for 'onlineSavingsMsg'
Learn More

TFA Dostmann Wireless Thermo-Hygrometer with 3 Transmitters KLIMA-at-HOME 2

Product Code. 17911662
Click to view available options
Humidity Range:
10 to 99%
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17911662

Brand: TFA Dostmann 30.3075.01

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

The wireless thermo-hygrometer with 3 transmitters KLIMA-at-HOME 2 is ideal for monitoring the indoor and outdoor climate in up to 4 locations. All monitored measuring points can be individually labeled on the station.

The Klima-at-Home 2 digital wireless thermo-hygrometer is supplied as a set with 3 thermo-hygro transmitters and is ideal for monitoring the indoor and outdoor climate in up to 4 locations. Indoors, monitoring the room temperature and humidity allows you to create a comfortable and healthy living climate through targeted ventilation and heating. This not only saves heating costs, but also contributes to general well-being. After all, whether we feel comfortable and cozy depends to a large extent on the quality of the indoor air. Mold and house dust mites only really multiply if the humidity is too high. But excessively dry indoor air can also affect our well-being in several ways. Skin and respiratory tracts are stressed and infections, allergies and asthma have an easy time of it. A room temperature of 18°C to 22°C and a relative humidity of 40 to 60% is ideal. Thanks to the maximum range of 100 m, the radio transmitter can also be set up outdoors and provide important values about the current outdoor climate. All monitored measuring points can be individually labeled on the station, making them easier to read. The time to the second is set automatically by radio signal, supplemented by the complete date or the day of the week. A 'feel-good' smiley shows you the current comfort level at the station's installation location. For all sensors, you can also call up the maximum and minimum values since the last reset as well as the current dew point at the station. The wireless thermometer Klima-at-Home 2 is battery-operated and can either be set up (the stand is removable) or mounted on the wall.

  • For the monitoring of indoor and outdoor climate
  • With internal sensor and 3 wireless radio-controlled transmitters (max. 100 m) for monitoring temperature and humidity
  • Max.-min.-function
  • Visual comfort zone
  • Dew point
  • Radio-controlled clock with date or weekday (6 languages)
  • Individual labeling of transmitter placements
  • Ideal for climate monitoring in up to 4 rooms

Specifications

Type Digital Thermohygrometer
Humidity Range 10 to 99%
Temperature Range (Metric) 0°C to 50°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.