Learn More
WTW™ Wireless Module For Plug Head Sensor
GREENER_CHOICE
Product Code. 15512016
Click to view available options
For Use With (Equipment):
WTS IDS Plug Head Sensors for Multiparameter Meters
Unit Size:
Each
Product Code. 15512016
Brand: WTW™ 108141
The Wireless Module For Plug Head Sensor is for use with the WTW™ range of IDS Intelligent Digital Sensors
  • Wireless modules for IDS plug head sensors are simply plugged onto the sensor and automatically connect to the meter, same module for all parameters
  • Safe and simple with no cables
  • Freedom to operate where a meter would usually be in the way
  • Even with portable field measurements under difficult conditions, one hand is always free
  • Recording measurement values by simply pressing a button on the sensor module
TRUSTED_SUSTAINABILITY
Specifications
Includes Re-chargeable LiPo battery 230 mAh
IP Rating Splash water protected to IP 66
For Use With (Equipment) WTS IDS Plug Head Sensors for Multiparameter Meters